siRNA Details
| Gene Symbol: Secisbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024002.1 | NP_076492.1 | 2 | 118 | ||
| XM_008771506.1 | XP_008769728.1 | 2 | 165 | |||
| GenBank mRNA | AJ251245.1 | - | 2 | 118 | ||
| FQ227169.1 | - | 2 | 138 | |||
| FQ231169.1 | - | 2 | 140 | |||
| FQ232320.1 | - | 2 | 146 | |||

Secisbp2
Genomic Map