siRNA Details
| Gene Symbol: Cfi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024157.1 | NP_077071.1 | 10 | 1023 | ||
| XM_008761526.1 | XP_008759748.1 | 11 | 1361 | |||
| GenBank mRNA | BC089798.1 | - | 10 | 1028 | 7321559 | |
| Y18965.1 | - | 10 | 1023 | |||

Cfi
Genomic Map