siRNA Details
| Gene Symbol: FICD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007076.2 | NP_009007.2 | 3 | 1129 | ||
| GenBank mRNA | AF049611.1 | - | 1 | 162 | ||
| AK000014.1 | - | 3 | 1103 | |||
| AK026074.1 | - | 3 | 1129 | |||
| AY358992.1 | - | 3 | 1098 | |||
| BC001342.1 | - | 3 | 1087 | 3462741 | ||

FICD
Genomic Map