siRNA Details
| Gene Symbol: Gnrhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031038.3 | NP_112300.2 | 1 | 78 | ||
| GenBank mRNA | CB775870.1 | - | 1 | 134 | ||
| L07646.1 | - | 1 | 64 | |||
| X68980.1 | - | 1 | 28 | |||
| X76635.1 | - | 1 | 17 | |||
More Information
Set Membership: |
|

Gnrhr
Genomic Map