siRNA Details
| Gene Symbol: Aldh6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031057.2 | NP_112319.2 | 3, 4 | 239 | ||
| XM_006240354.2 | XP_006240416.1 | 3, 4 | 604 | |||
| XM_008764862.1 | XP_008763084.1 | 2, 3 | 143 | |||
| XM_008764863.1 | XP_008763085.1 | 3, 4 | 241 | |||
| GenBank mRNA | CB760062.1 | - | 3, 4 | 296 | ||
| DN948946.1 | - | 3, 4 | 236 | |||

Aldh6a1
Genomic Map