siRNA Details
| Gene Symbol: Aldh6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031057.2 | NP_112319.2 | 9 | 1257 | ||
| XM_006240354.2 | XP_006240416.1 | 9 | 1622 | |||
| XM_008764862.1 | XP_008763084.1 | 8 | 1161 | |||
| XM_008764863.1 | XP_008763085.1 | 9 | 1259 | |||

Aldh6a1
Genomic Map