siRNA Details
| Gene Symbol: Pggt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031082.1 | NP_112344.1 | 2 | 227 | ||
| XM_006254691.2 | XP_006254753.1 | 2 | 1467 | |||
| XM_008772177.1 | XP_008770399.1 | 3 | 229 | |||
| GenBank mRNA | L24116.1 | - | 2 | 227 | ||

Pggt1b
Genomic Map