siRNA Details
| Gene Symbol: Rock1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031098.1 | NP_112360.1 | 3 | 334 | ||
| XM_006254400.2 | XP_006254462.1 | 3 | 1086 | |||
| XM_006254401.2 | XP_006254463.1 | 3 | 1086 | |||
| XM_008771957.1 | XP_008770179.1 | 3 | 1086 | |||
| GenBank mRNA | FQ225745.1 | - | 4 | 810 | ||
| FQ226663.1 | - | 1 | 40 | |||
| U61266.1 | - | 3 | 334 | |||

Rock1
Genomic Map