siRNA Details
Gene Symbol: Ssr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031120.2 | NP_112382.2 | 2 | 704 | ||
GenBank mRNA | AY724496.1 | - | 2 | 707 | ||
BC062015.1 | - | 2 | 704 | 5621495 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ssr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031120.2 | NP_112382.2 | 2 | 704 | ||
GenBank mRNA | AY724496.1 | - | 2 | 707 | ||
BC062015.1 | - | 2 | 704 | 5621495 |