siRNA Details
| Gene Symbol: Ube2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031138.2 | NP_112400.1 | 6 | 485 | ||
| XM_006246343.2 | XP_006246405.1 | 7 | 510 | |||
| GenBank mRNA | BC070946.1 | - | 6 | 478 | 7103580 | |
| DN931313.1 | - | 6 | 515 | |||
| FQ216310.1 | - | 4 | 265 | |||
| FQ216742.1 | - | 3 | 204 | |||
| FQ222428.1 | - | 6 | 502 | |||
| FQ229894.1 | - | 6 | 496 | |||
| M62388.1 | - | 6 | 427 | |||
| Gene Symbol: LOC103694902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773893.1 | XP_008772115.1 | 7 | 509 | ||

Ube2b
Genomic Map