siRNA Details
| Gene Symbol: Psmc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031149.1 | NP_112411.1 | 9 | 952 | ||
| GenBank mRNA | AB000491.1 | - | 9 | 952 | ||
| BC058462.1 | - | 9 | 948 | 6891005 | ||
| D83521.1 | - | 9 | 970 | |||
| FQ213263.1 | - | 9 | 969 | |||
| FQ213735.1 | - | 9 | 944 | |||

Psmc5
Genomic Map