siRNA Details
| Gene Symbol: Sh3gl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031238.2 | NP_112517.1 | 3, 4 | 533 | ||
| XM_006229480.2 | XP_006229542.1 | 3, 4 | 321 | |||
| XM_006229481.2 | XP_006229543.1 | 3, 4 | 414 | |||
| XM_006229483.2 | XP_006229545.1 | 3, 4 | 520 | |||
| GenBank mRNA | AF009604.1 | - | 1, 2 | 17 | ||
| AF227439.1 | - | 3, 4 | 536 | |||
| BC078800.1 | - | 3, 4 | 536 | 7135194 | ||

Sh3gl3
Genomic Map