siRNA Details
| Gene Symbol: Sh3gl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031239.2 | NP_112518.1 | 4 | 347 | ||
| GenBank mRNA | AB008161.1 | - | 4 | 347 | ||
| AF009602.1 | - | 4 | 332 | |||
| BC070893.1 | - | 4 | 343 | 7099223 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh3gl1| Gene Symbol: Sh3gl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031239.2 | NP_112518.1 | 4 | 347 | ||
| GenBank mRNA | AB008161.1 | - | 4 | 347 | ||
| AF009602.1 | - | 4 | 332 | |||
| BC070893.1 | - | 4 | 343 | 7099223 | ||