siRNA Details
| Gene Symbol: Polr2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031335.3 | NP_112625.1 | 3 | 197 | ||
| GenBank mRNA | AB017711.1 | - | 3 | 197 | ||
| BC083552.1 | - | 3 | 237 | 7190444 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Polr2f| Gene Symbol: Polr2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031335.3 | NP_112625.1 | 3 | 197 | ||
| GenBank mRNA | AB017711.1 | - | 3 | 197 | ||
| BC083552.1 | - | 3 | 237 | 7190444 | ||