siRNA Details
| Gene Symbol: Scn7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031686.1 | NP_113874.1 | 1 | 136 | ||
| GenBank mRNA | Y09164.1 | - | 1 | 136 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn7a| Gene Symbol: Scn7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031686.1 | NP_113874.1 | 1 | 136 | ||
| GenBank mRNA | Y09164.1 | - | 1 | 136 | ||