siRNA Details
| Gene Symbol: Nefl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031783.1 | NP_113971.1 | 1 | 733 | ||
| GenBank mRNA | AF031880.1 | - | 1 | 733 | ||
| M25638.1 | - | 1 | 71 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nefl| Gene Symbol: Nefl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031783.1 | NP_113971.1 | 1 | 733 | ||
| GenBank mRNA | AF031880.1 | - | 1 | 733 | ||
| M25638.1 | - | 1 | 71 | |||