siRNA Details
| Gene Symbol: Rest | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031788.1 | NP_113976.1 | 1 | 190 | ||
| XM_006250847.2 | XP_006250909.1 | 269 | ||||
| XM_006250848.1 | XP_006250910.1 | 2 | 274 | |||
| XM_006250849.2 | XP_006250911.1 | 2 | 532 | |||
| GenBank mRNA | AF037199.1 | - | 1 | 190 | ||
| AF037203.1 | - | 1 | 190 | |||

Rest
Genomic Map