siRNA Details
| Gene Symbol: Gabbr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031802.1 | NP_113990.1 | 2 | 512 | ||
| GenBank mRNA | AF058795.1 | - | 2 | 512 | ||
| AF074482.1 | - | 2 | 362 | |||
| AF109405.1 | - | 2 | 479 | |||
More Information
Set Membership: |
|

Gabbr2
Genomic Map