siRNA Details
| Gene Symbol: Cngb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031809.1 | NP_113997.1 | 8, 9 | 687 | ||
| XM_006255124.2 | XP_006255186.1 | 8, 9 | 748 | |||
| XM_006255125.2 | XP_006255187.1 | 8, 9 | 738 | |||
| GenBank mRNA | AJ000496.1 | - | 8, 9 | 687 | ||
| AJ224680.1 | - | 8, 9 | 687 | |||

Cngb1
Genomic Map