siRNA Details
| Gene Symbol: Taldo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031811.3 | NP_113999.2 | 4 | 390 | ||
| GenBank mRNA | AF069306.1 | - | 4 | 376 | ||
| BC059126.1 | - | 4 | 394 | 6889440 | ||
| FQ213939.1 | - | 4 | 402 | |||
| FQ219168.1 | - | 4 | 416 | |||

Taldo1
Genomic Map