siRNA Details
| Gene Symbol: Taldo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031811.3 | NP_113999.2 | 8 | 1138 | ||
| GenBank mRNA | BC059126.1 | - | 8 | 1142 | 6889440 | |
| FQ213939.1 | - | 8 | 1150 | |||
| FQ219168.1 | - | 8 | 1164 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Taldo1| Gene Symbol: Taldo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031811.3 | NP_113999.2 | 8 | 1138 | ||
| GenBank mRNA | BC059126.1 | - | 8 | 1142 | 6889440 | |
| FQ213939.1 | - | 8 | 1150 | |||
| FQ219168.1 | - | 8 | 1164 | |||