siRNA Details
| Gene Symbol: Rps6kb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031985.1 | NP_114191.1 | 15 | 1800 | ||
| GenBank mRNA | BC089789.1 | - | 16 | 1870 | ||
| M58340.1 | - | 15 | 1800 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rps6kb1| Gene Symbol: Rps6kb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031985.1 | NP_114191.1 | 15 | 1800 | ||
| GenBank mRNA | BC089789.1 | - | 16 | 1870 | ||
| M58340.1 | - | 15 | 1800 | |||