siRNA Details
| Gene Symbol: Eif2b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032058.2 | NP_114447.2 | 3, 4 | 467 | ||
| GenBank mRNA | BC070506.1 | - | 3, 4 | 467 | 7097686 | |
| FQ232990.1 | - | 3, 4 | 476 | |||
| U31880.1 | - | 3, 4 | 470 | |||
| U83914.1 | - | 3, 4 | 438 | |||

Eif2b2
Genomic Map