siRNA Details
| Gene Symbol: Kalrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032062.2 | NP_114451.2 | 31 | 4726 | ||
| GenBank mRNA | AF229255.1 | - | 22 | 3387 | ||
| AF230644.1 | - | 31 | 4726 | |||
| AF232668.1 | - | 30 | 4696 | |||
| AF232669.1 | - | 30 | 4696 | |||
| U88156.1 | - | 5079 | ||||
| U88157.1 | - | 30 | 4696 | |||

Kalrn
Genomic Map