siRNA Details
| Gene Symbol: Hmga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032070.1 | NP_114459.1 | 3 | 629 | ||
| GenBank mRNA | U89695.1 | - | 3 | 629 | ||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: Hmga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032070.1 | NP_114459.1 | 3 | 629 | ||
| GenBank mRNA | U89695.1 | - | 3 | 629 | ||
Hmga2
Genomic Map