siRNA Details
Gene Symbol: Chn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032084.1 | NP_114473.1 | 5 | 410 | ||
GenBank mRNA | L07494.1 | - | 5 | 410 | ||
X69489.1 | - | 5 | 410 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Chn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032084.1 | NP_114473.1 | 5 | 410 | ||
GenBank mRNA | L07494.1 | - | 5 | 410 | ||
X69489.1 | - | 5 | 410 |