siRNA Details
| Gene Symbol: Chn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032084.1 | NP_114473.1 | 9 | 911 | ||
| GenBank mRNA | L07494.1 | - | 9 | 911 | ||
| X69489.1 | - | 9 | 911 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chn2| Gene Symbol: Chn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032084.1 | NP_114473.1 | 9 | 911 | ||
| GenBank mRNA | L07494.1 | - | 9 | 911 | ||
| X69489.1 | - | 9 | 911 | |||