siRNA Details
| Gene Symbol: Rims1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052829.1 | NP_439894.1 | 6 | 992 | ||
| XM_008766936.1 | XP_008765158.1 | 6 | 992 | |||
| GenBank mRNA | AF007836.1 | - | 6 | 992 | ||
| AF199333.1 | - | 6 | 992 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rims1| Gene Symbol: Rims1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052829.1 | NP_439894.1 | 6 | 992 | ||
| XM_008766936.1 | XP_008765158.1 | 6 | 992 | |||
| GenBank mRNA | AF007836.1 | - | 6 | 992 | ||
| AF199333.1 | - | 6 | 992 | |||