siRNA Details
| Gene Symbol: Homer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053310.1 | NP_445762.1 | 3 | 249 | ||
| XM_006252890.2 | XP_006252952.1 | 4 | 375 | |||
| XM_006252891.2 | XP_006252953.1 | 4 | 372 | |||
| XM_008771094.1 | XP_008769316.1 | 4 | 373 | |||
| XM_008771095.1 | XP_008769317.1 | 4 | 373 | |||
| GenBank mRNA | AB020879.2 | - | 3 | 249 | ||
| BC070922.1 | - | 4 | 358 | |||

Homer3
Genomic Map