siRNA Details
| Gene Symbol: Agps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053350.2 | NP_445802.2 | 1, 2 | 319 | ||
| GenBank mRNA | AF121052.1 | - | 1, 2 | 224 | ||
| AJ238006.1 | - | 1, 2 | 224 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Agps| Gene Symbol: Agps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053350.2 | NP_445802.2 | 1, 2 | 319 | ||
| GenBank mRNA | AF121052.1 | - | 1, 2 | 224 | ||
| AJ238006.1 | - | 1, 2 | 224 | |||