siRNA Details
| Gene Symbol: Ackr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053352.1 | NP_445804.1 | 2 | 438 | ||
| XM_006245479.2 | XP_006245541.1 | 3 | 632 | |||
| XM_008767299.1 | XP_008765521.1 | 2 | 3757 | |||
| XM_008767300.1 | XP_008765522.1 | 3 | 2718 | |||
| GenBank mRNA | AF118816.1 | - | 2 | 438 | ||
| AJ010828.1 | - | 2 | 433 | |||

Ackr3
Genomic Map