siRNA Details
| Gene Symbol: Cd40lg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053353.1 | NP_445805.1 | 2 | 212 | ||
| GenBank mRNA | AF013985.1 | - | 2 | 212 | ||
| AF116582.1 | - | 2 | 212 | |||
| EF066490.1 | - | 2 | 212 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd40lg| Gene Symbol: Cd40lg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053353.1 | NP_445805.1 | 2 | 212 | ||
| GenBank mRNA | AF013985.1 | - | 2 | 212 | ||
| AF116582.1 | - | 2 | 212 | |||
| EF066490.1 | - | 2 | 212 | |||