siRNA Details
| Gene Symbol: Slpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053372.1 | NP_445824.1 | 4 | 447 | ||
| GenBank mRNA | AF151982.1 | - | 4 | 447 | ||
| AF178426.1 | - | 4 | 447 | |||
| AF421377.1 | - | 4 | 457 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slpi| Gene Symbol: Slpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053372.1 | NP_445824.1 | 4 | 447 | ||
| GenBank mRNA | AF151982.1 | - | 4 | 447 | ||
| AF178426.1 | - | 4 | 447 | |||
| AF421377.1 | - | 4 | 457 | |||