siRNA Details
Gene Symbol: Cd93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053383.1 | NP_445835.1 | 1 | 346 | ||
GenBank mRNA | AF136537.1 | - | 1 | 273 | ||
AF160978.1 | - | 1 | 346 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053383.1 | NP_445835.1 | 1 | 346 | ||
GenBank mRNA | AF136537.1 | - | 1 | 273 | ||
AF160978.1 | - | 1 | 346 |