siRNA Details
| Gene Symbol: Cd93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053383.1 | NP_445835.1 | 1 | 351 | ||
| GenBank mRNA | AF136537.1 | - | 1 | 278 | ||
| AF160978.1 | - | 1 | 351 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd93| Gene Symbol: Cd93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053383.1 | NP_445835.1 | 1 | 351 | ||
| GenBank mRNA | AF136537.1 | - | 1 | 278 | ||
| AF160978.1 | - | 1 | 351 | |||