siRNA Details
| Gene Symbol: ZNF79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286696.1 | NP_001273625.1 | 5 | 2087 | ||
| NM_001286697.1 | NP_001273626.1 | 6 | 2089 | |||
| NM_001286698.1 | NP_001273627.1 | 3 | 1864 | |||
| NM_001322260.1 | NP_001309189.1 | 6 | 2218 | |||
| NM_007135.2 | NP_009066.2 | 5 | 1980 | |||
| XM_006717279.3 | XP_006717342.1 | 6 | 2188 | |||
| GenBank mRNA | AK054606.1 | - | 5 | 1972 | ||
| BC062309.1 | - | 5 | 1935 | 3944657 | ||

ZNF79
Genomic Map