siRNA Details
Gene Symbol: Il12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053390.1 | NP_445842.1 | 6, 7 | 525 | ||
GenBank mRNA | AF177031.1 | - | 6, 7 | 525 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Il12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053390.1 | NP_445842.1 | 6, 7 | 525 | ||
GenBank mRNA | AF177031.1 | - | 6, 7 | 525 |