siRNA Details
Gene Symbol: Cxcr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053415.1 | NP_445867.1 | 2 | 252 | ||
GenBank mRNA | AF223642.1 | - | 2 | 252 | ||
BC091136.1 | - | 1 | 107 | 7376065 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cxcr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053415.1 | NP_445867.1 | 2 | 252 | ||
GenBank mRNA | AF223642.1 | - | 2 | 252 | ||
BC091136.1 | - | 1 | 107 | 7376065 |