siRNA Details
| Gene Symbol: Slc17a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053427.1 | NP_445879.1 | 12 | 3270 | ||
| XM_006229265.1 | XP_006229327.1 | 12 | 2799 | |||
| GenBank mRNA | AF271235.1 | - | 12 | 3270 | ||
| FQ211915.1 | - | 1 | 103 | |||

Slc17a6
Genomic Map