siRNA Details
| Gene Symbol: Stau1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109907.1 | NP_001103377.1 | 3, 4 | 486 | ||
| NM_053436.2 | NP_445888.1 | 3, 4 | 486 | |||
| GenBank mRNA | AF227200.1 | - | 3, 4 | 473 | ||
| AF290989.1 | - | 3, 4 | 473 | |||
| AJ010200.1 | - | 1, 2 | 167 | |||
| BC081755.1 | - | 3, 4 | 444 | |||
| BC101858.1 | - | 3, 4 | 482 | 7937343 | ||
| DN932990.1 | - | 3, 4 | 515 | |||

Stau1
Genomic Map