siRNA Details
| Gene Symbol: Cnga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053497.1 | NP_445949.1 | 1, 2 | 115 | ||
| GenBank mRNA | U93851.1 | - | 1, 2 | 115 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnga1| Gene Symbol: Cnga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053497.1 | NP_445949.1 | 1, 2 | 115 | ||
| GenBank mRNA | U93851.1 | - | 1, 2 | 115 | ||