siRNA Details
| Gene Symbol: Mterf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053499.1 | NP_445951.1 | 3 | 935 | ||
| XM_006236069.2 | XP_006236131.1 | 4 | 1050 | |||
| XM_006236070.2 | XP_006236132.1 | 4 | 1047 | |||
| XM_006236071.2 | XP_006236133.1 | 3 | 922 | |||
| XM_006236072.2 | XP_006236134.1 | 3 | 919 | |||
| XM_006236073.2 | XP_006236135.1 | 2 | 873 | |||
| GenBank mRNA | AJ292524.1 | - | 3 | 935 | ||

Mterf
Genomic Map