siRNA Details
| Gene Symbol: Cdhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053572.1 | NP_446024.1 | 17 | 3278 | ||
| GenBank mRNA | AB053449.1 | - | 17 | 3278 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdhr1| Gene Symbol: Cdhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053572.1 | NP_446024.1 | 17 | 3278 | ||
| GenBank mRNA | AB053449.1 | - | 17 | 3278 | ||