siRNA Details
| Gene Symbol: Cabin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053575.2 | NP_446027.2 | 4, 5 | 543 | ||
| GenBank mRNA | AF061947.1 | - | 4, 5 | 549 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cabin1| Gene Symbol: Cabin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053575.2 | NP_446027.2 | 4, 5 | 543 | ||
| GenBank mRNA | AF061947.1 | - | 4, 5 | 549 | ||