siRNA Details
| Gene Symbol: Zfp423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053583.2 | NP_446035.2 | 585 | |||
| GenBank mRNA | L03386.1 | - | 489 | |||
| U92564.1 | - | 583 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp423| Gene Symbol: Zfp423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053583.2 | NP_446035.2 | 585 | |||
| GenBank mRNA | L03386.1 | - | 489 | |||
| U92564.1 | - | 583 | ||||