siRNA Details
| Gene Symbol: Dut | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040271.1 | NP_001035361.1 | 6 | 562 | ||
| NM_053592.2 | NP_446044.1 | 7 | 682 | |||
| XM_008762188.1 | XP_008760410.1 | 6 | 649 | |||
| GenBank mRNA | CB325379.1 | - | 6 | 576 | ||
| FQ220451.1 | - | 717 | ||||
| FQ233848.1 | - | 2 | 182 | |||
| U64030.2 | - | 7 | 682 | |||

Dut
Genomic Map