siRNA Details
| Gene Symbol: Magi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053621.1 | NP_446073.1 | 3, 4 | 688 | ||
| GenBank mRNA | AF034863.1 | - | 3, 4 | 688 | ||
| AF130819.1 | - | 2, 3 | 475 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Magi2| Gene Symbol: Magi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053621.1 | NP_446073.1 | 3, 4 | 688 | ||
| GenBank mRNA | AF034863.1 | - | 3, 4 | 688 | ||
| AF130819.1 | - | 2, 3 | 475 | |||