siRNA Details
| Gene Symbol: Gfm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053625.1 | NP_446077.1 | 1, 2 | 225 | ||
| GenBank mRNA | BC085721.1 | - | 1, 2 | 181 | 7192461 | |
| L14684.1 | - | 1, 2 | 225 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gfm1| Gene Symbol: Gfm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053625.1 | NP_446077.1 | 1, 2 | 225 | ||
| GenBank mRNA | BC085721.1 | - | 1, 2 | 181 | 7192461 | |
| L14684.1 | - | 1, 2 | 225 | |||