siRNA Details
| Gene Symbol: Cdc42bpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053657.1 | NP_446109.1 | 5 | 1037 | ||
| GenBank mRNA | AF021935.1 | - | 5 | 1037 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc42bpa| Gene Symbol: Cdc42bpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053657.1 | NP_446109.1 | 5 | 1037 | ||
| GenBank mRNA | AF021935.1 | - | 5 | 1037 | ||