siRNA Details
| Gene Symbol: Phyh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053674.1 | NP_446126.1 | 5 | 463 | ||
| GenBank mRNA | AF121345.1 | - | 5 | 463 | ||
| BC086573.1 | - | 5 | 458 | 7306754 | ||
| FQ216943.1 | - | 3 | 295 | |||
| FQ224924.1 | - | 3 | 295 | |||

Phyh
Genomic Map